accessibility menu, dialog, popup

UserName

CAS RN 85107-54-6

HOBOHNOOOHO

CAS RN 85107-54-6

IUPAC Name: 4-(dihydroxyboranyl)-3-nitrobenzoic acid
Synonyms: 4-(dihydroxyboranyl)-3-nitrobenzoic acid
Molecular Weight (g/mol): 210.94
Molecular Formula: C7H6BNO6
InChi Key: APBXPKJLXBBPQJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results