accessibility menu, dialog, popup

UserName

CAS RN 850568-13-7

BONHCH3CH3OHHO

CAS RN 850568-13-7

IUPAC Name: {4-[(2-methylpropyl)carbamoyl]phenyl}boronic acid
Synonyms: 4-[(2-methylpropyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 221.06
Molecular Formula: C11H16BNO3
InChi Key: DUBSEAOPNSRHHN-UHFFFAOYSA-N
SMILES: CC(C)CNC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results