accessibility menu, dialog, popup

UserName

CAS RN 84966-28-9

OON

CAS RN 84966-28-9

IUPAC Name: 5-(pyrrolidin-1-yl)furan-2-carbaldehyde
Synonyms: 5-(pyrrolidin-1-yl)furan-2-carbaldehyde
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: TZLGFEAULCWLDT-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(O1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results