CAS RN 823218-99-1
IUPAC Name:
5-(2,4,6-trimethylphenyl)-3-oxa-4-azatricyclo[5.2.1.0²,⁶]dec-4-ene
Synonyms:
5-(2,4,6-trimethylphenyl)-3-oxa-4-azatricyclo[5.2.1.0²,⁶]dec-4-ene
Molecular Weight (g/mol):
255.36
Molecular Formula:
C17H21NO
InChi Key:
WKLZNTYMDOPBSE-UHFFFAOYNA-N
SMILES:
CC1=CC(C)=C(C2=NOC3C4CCC(C4)C23)C(C)=C1
1
–
2
of
2
results
1
SN 2, MedChemExpress
MedChemExpress SN 2 is a potent activator of TRPML3 ion channel with an EC50 of 1.8 μM. SN 2 also acts as a potent inhibitor of Dengue virus 2 (DENV2) and Zika virus (ZIKV).
2
SN 2, Tocris Bioscience™
CAS: 823218-99-1 Molecular Formula: C17H21NO Molecular Weight (g/mol): 255.361 InChI Key: WKLZNTYMDOPBSE-UHFFFAOYSA-N Synonym: sn 2,sn-2,5-mesityl-3-oxa-4-azatricyclo 5.2.1.0∼2,6∼ dec-4-ene,5-mesityl-3-oxa-4-azatricyclo 5.2.1.02,6 dec-4-ene,d0g1ln,5-mesityl-3-oxa-4-azatricyclo 5.2.1.0 2,6 dec-4-ene,3a,4,5,6,7,7a-hexahydro-3-2,4,6-trimethylphenyl-4,7-methano-1,2-benzisoxazole PubChem CID: 11834987 SMILES: CC1=CC(=C(C(=C1)C)C2=NOC3C2C4CCC3C4)C