accessibility menu, dialog, popup

UserName

CAS RN 81-23-2

H3C(R)OHO(R)(S)(R)(S)O(R)CH3(S)CH3O(S)O

CAS RN 81-23-2

IUPAC Name: (4R)-4-[(1R,3aS,3bR,5aS,9aS,9bS,11aR)-9a,11a-dimethyl-4,7,11-trioxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: dehydrocholic dehydrocholic acid, lithium salt ketochol chologon oxycholin ketocholanic acid ciba vision brand of dehydrocholic acid deidrocolico vita dehydro-cholic acid decholin
Molecular Weight (g/mol): 402.53
Molecular Formula: C24H34O5
InChi Key: OHXPGWPVLFPUSM-KLRNGDHRSA-N
SMILES: C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]1(C)CCC(=O)C[C@H]1CC3=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results