accessibility menu, dialog, popup

UserName

OHNNHON(Z)

CAS RN 796-42-9

IUPAC Name: N'-{[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}pyridine-4-carbohydrazide
Synonyms: N'-{[(1Z)-2-oxonaphthalen-1-ylidene]methyl}pyridine-4-carbohydrazide
Molecular Weight (g/mol): 291.31
Molecular Formula: C17H13N3O2
InChi Key: UBECDIQQXGETMR-PTNGSMBKSA-N
SMILES: O=C(NN\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=NC=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)