accessibility menu, dialog, popup

UserName

CAS RN 7789-92-6

H3COOCH3OCH3

CAS RN 7789-92-6

IUPAC Name: 1,1,3-triethoxypropane
Synonyms: propane, 1,3,3-triethoxy-
Molecular Weight (g/mol): 176.26
Molecular Formula: C9H20O3
InChi Key: LGICWIVABSMSDK-UHFFFAOYSA-N
SMILES: CCOCCC(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results