accessibility menu, dialog, popup

UserName

CAS RN 771573-34-3

SFFFFFNH2

CAS RN 771573-34-3

IUPAC Name: 1-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]methanamine
Synonyms: 1-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]methanamine
Molecular Weight (g/mol): 233.2
Molecular Formula: C7H8F5NS
InChi Key: FJOVSWPDVAFUMX-UHFFFAOYSA-N
SMILES: NCC1=CC(=CC=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results