CAS RN 7704-67-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Erythromycin Thiocyanate, TRC
CAS: 7704-67-8 Molecular Formula: C37H67NO13 • CHNS Molecular Weight (g/mol): 733.935898 Synonym: Bactenix V 300,Erythrocin Thiocyanate,Erytrovet,Gallimycin 10,Gallimycin 50 Pro-Gallimycin 10; SMILES: CC[C@@H]1[C@](C)([C@@H]([C@@H](C)C(=O)[C@H](C)C[C@](C)([C@@H]([C@@H](C)[C@@H]([C@@H](C)C(=O)O1)O[C@H]2C[C@](C)([C@H]([C@H](C)O2)O)OC)O[C@H]3[C@@H]([C@H](C[C@@H](C)O3)N(C)C)O)O)O)O.C(#N)S