CAS RN 7691-02-3
CAS RN 7691-02-3
IUPAC Name:
bis(ethenyldimethylsilyl)amine
Synonyms:
bis(ethenyldimethylsilyl)amine
Molecular Weight (g/mol):
185.42
Molecular Formula:
C8H19NSi2
InChi Key:
WYUIWUCVZCRTRH-UHFFFAOYSA-N
SMILES:
C[Si](C)(N[Si](C)(C)C=C)C=C
1
–
1
of
1
results
1
1,1,3,3-Tetramethyl-1,3-divinyldisilazane, 96%, Thermo Scientific™
CAS: 7691-02-3 MDL Number: MFCD00042846