accessibility menu, dialog, popup

UserName

CAS RN 76393-18-5

BrNCOBrF

CAS RN 76393-18-5

IUPAC Name: 1,3-dibromo-5-fluoro-2-isocyanatobenzene
Synonyms: 1,3-dibromo-5-fluoro-2-isocyanatobenzene
Molecular Weight (g/mol): 294.91
Molecular Formula: C7H2Br2FNO
InChi Key: NYPBOWHIPOBXBA-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(N=C=O)C(Br)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results