accessibility menu, dialog, popup

UserName

CAS RN 76008-73-6

H3COOBrCl

CAS RN 76008-73-6

IUPAC Name: ethyl 5-bromo-2-chlorobenzoate
Synonyms: ethyl 5-bromo-2-chlorobenzoate
Molecular Weight (g/mol): 263.52
Molecular Formula: C9H8BrClO2
InChi Key: AMGWDYLEMSMUIO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(Br)=CC=C1Cl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)