accessibility menu, dialog, popup

UserName

CAS RN 75-00-3

H3CCl

CAS RN 75-00-3

IUPAC Name: chloroethane
Synonyms: ethyl chloride muriatic ether chlorure d'ethyle chloroaethan chelen ethyl chloride preparation (product) hydrochloric ether kelene gebauer brand of ethyl chloride etylu chlorek
Molecular Weight (g/mol): 64.51
Molecular Formula: C2H5Cl
InChi Key: HRYZWHHZPQKTII-UHFFFAOYSA-N
SMILES: CCCl
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results