accessibility menu, dialog, popup

UserName

CH3OHNO

CAS RN 74588-78-6

IUPAC Name: 2-benzoyl-5-methoxy-1H-indole
Synonyms: 2-benzoyl-5-methoxy-1H-indole
Molecular Weight (g/mol): 251.29
Molecular Formula: C16H13NO2
InChi Key: ICMIJSRDISNKOC-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=CC2=C1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results