accessibility menu, dialog, popup

UserName

CAS RN 745784-10-5

HOBOHNF

CAS RN 745784-10-5

IUPAC Name: (2-fluoroquinolin-3-yl)boronic acid
Synonyms: 2-fluoroquinolin-3-ylboronic acid
Molecular Weight (g/mol): 190.97
Molecular Formula: C9H7BFNO2
InChi Key: SAUJOURLTDSVOK-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)N=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results