accessibility menu, dialog, popup

UserName

CAS RN 745784-05-8

IUPAC Name: (2-bromoquinolin-3-yl)boronic acid
Synonyms: 2-bromoquinolin-3-ylboronic acid
Molecular Weight (g/mol): 251.87
Molecular Formula: C9H7BBrNO2
InChi Key: RUDILYIFXMOEIO-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Br)N=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results