accessibility menu, dialog, popup

UserName

CAS RN 74483-46-8

CH3FFFCl

CAS RN 74483-46-8

IUPAC Name: 2-chloro-4-methyl-1-(trifluoromethyl)benzene
Synonyms: 2-chloro-4-methyl-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.58
Molecular Formula: C8H6ClF3
InChi Key: OHDYNLHQHOFWGR-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results