accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 7441-43-2

IUPAC Name: 1-(4-ethylphenyl)methanamine
Synonyms: 1-(4-ethylphenyl)methanamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: DGAGEFUEKIORSQ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CN)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results