accessibility menu, dialog, popup

UserName

CAS RN 7435-83-8

CH3CH3CH3ClH3C

CAS RN 7435-83-8

IUPAC Name: 3-(chloromethyl)-1,2,4,5-tetramethylbenzene
Synonyms: 3-(chloromethyl)-1,2,4,5-tetramethylbenzene
Molecular Weight (g/mol): 182.69
Molecular Formula: C11H15Cl
InChi Key: UGAPPXGBBWAIGT-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(CCl)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results