CAS RN 74-97-5
CAS RN 74-97-5
IUPAC Name:
bromo(chloro)methane
Synonyms:
bromochloromethane fluorocarbon 1011 ccris 817 C-H2-br-cl methylene chlorobromide
Molecular Weight (g/mol):
129.38
Molecular Formula:
CH2BrCl
InChi Key:
JPOXNPPZZKNXOV-UHFFFAOYSA-N
SMILES:
ClCBr