accessibility menu, dialog, popup

UserName

CH3HS

CAS RN 74-93-1

IUPAC Name: methanethiol
Synonyms: methanethiol mercury (2+) salt methylmercaptan methyl mercaptan (natural) methylmercaptan, mercury (2+) salt methvtiolo sodium salt methylmercaptan metilmercaptano rcra waste number U153 methyl mercaptan (substance) methylmercaptan, lead (2+) salt
Molecular Weight (g/mol): 48.1
Molecular Formula: CH4S
InChi Key: LSDPWZHWYPCBBB-UHFFFAOYSA-N
SMILES: CS
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results