accessibility menu, dialog, popup

UserName

CAS RN 737751-95-0

OOHNH2Br

CAS RN 737751-95-0

IUPAC Name: 3-amino-3-(2-bromophenyl)propanoic acid
Synonyms: 3-amino-3-(2-bromophenyl)propanoic acid
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: OETRFEPZCAGEMK-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)