accessibility menu, dialog, popup

UserName

CAS RN 737000-91-8

ONCS

CAS RN 737000-91-8

IUPAC Name: {[(2-isothiocyanatocyclopentyl)oxy]methyl}benzene
Synonyms: {[(2-isothiocyanatocyclopentyl)oxy]methyl}benzene
Molecular Weight (g/mol): 233.33
Molecular Formula: C13H15NOS
InChi Key: XPXSISUCDSDZAB-UHFFFAOYNA-N
SMILES: S=C=NC1CCCC1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results