CAS RN 73509-20-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Indole-d7, TRC
CAS: 73509-20-3 Molecular Formula: C8 D7 N Molecular Weight (g/mol): 124.19 Synonym: 1H-Indole-1,2,3,4,5,6,7-d7 (9CI, ACI),Indole-d7,Indole-D7 IUPAC Name: 1,2,3,4,5,6,7-heptadeuterioindole SMILES: [2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])n2[2H]
2
Indole-d7, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.