accessibility menu, dialog, popup

UserName

CAS RN 7335-17-3

H3CCH3OCH3CH3

CAS RN 7335-17-3

IUPAC Name: 3,5-dimethyloctan-4-one
Synonyms: 3,5-dimethyloctan-4-one
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: HIYIOWIWNUWNKQ-UHFFFAOYNA-N
SMILES: CCCC(C)C(=O)C(C)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)