accessibility menu, dialog, popup

UserName

CAS RN 71989-43-0

H3C(R)O(S)NH2OOH3CCH3CH3CH3HCl

CAS RN 71989-43-0

IUPAC Name: methyl 2-amino-3-(tert-butoxy)butanoate hydrochloride
Synonyms: methyl 2-amino-3-(tert-butoxy)butanoate hydrochloride
Molecular Weight (g/mol): 225.71
Molecular Formula: C9H20ClNO3
InChi Key: SDHKEUUZUMQSAD-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(N)C(C)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)