accessibility menu, dialog, popup

UserName

CAS RN 71701-92-3

IUPAC Name: 3-bromo-2-chloro-5-(trifluoromethyl)pyridine
Synonyms: 3-bromo-2-chloro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 260.44
Molecular Formula: C6H2BrClF3N
InChi Key: GVQLMGPWTHAUPV-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=C(Cl)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results