accessibility menu, dialog, popup

UserName

CAS RN 701-83-7

CH3OOF

CAS RN 701-83-7

IUPAC Name: 3-fluorophenyl acetate
Synonyms: 3-fluorophenyl acetate
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: IAWZWMGUTKRLQB-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC(F)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)