accessibility menu, dialog, popup

UserName

CAS RN 699-12-7

HOS

CAS RN 699-12-7

IUPAC Name: 2-(phenylsulfanyl)ethan-1-ol
Synonyms: 2-(phenylsulfanyl)ethanol
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: KWWZHCSQVRVQGF-UHFFFAOYSA-N
SMILES: OCCSC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)