CAS RN 6968-76-9
IUPAC Name:
1-(3-methoxyphenyl)piperazine hydrochloridyl
Synonyms:
1-(3-methoxyphenyl)piperazine hydrochloridyl
Molecular Weight (g/mol):
227.71
Molecular Formula:
C11H16ClN2O
InChi Key:
UYZISVLMMYHXMS-UHFFFAOYSA-N
SMILES:
[Cl].COC1=CC=CC(=C1)N1CCNCC1
1
–
2
of
2
results
2
1-(3-Methoxyphenyl)piperazine dihydrochloride, 97%, Thermo Scientific™
CAS: 6968-76-9 Molecular Formula: C11H18Cl2N2O Molecular Weight (g/mol): 265.178 MDL Number: MFCD00035285 InChI Key: UKUNKQNESKRETR-UHFFFAOYSA-N Synonym: 1-3-methoxyphenyl piperazine dihydrochloride,unii-q0t82qhg7n,1-3-methoxyphenyl piperazine 2hcl,q0t82qhg7n,piperazine, 1-3-methoxyphenyl-, dihydrochloride,1-3-methoxyphenyl piperazindihydrochlorid,1-3-methoxyphenyl piperazine hcl,pubchem15280,1-3-methoxyphenyl piperazinehydrochloride,n-3-methoxyphenyl piperazine dihydrochloride PubChem CID: 81429 IUPAC Name: 1-(3-methoxyphenyl)piperazine;dihydrochloride SMILES: COC1=CC=CC(=C1)N2CCNCC2.Cl.Cl