CAS RN 69363-14-0
CAS RN 69363-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Schisanhenol, MedChemExpress
MedChemExpress Schisanhenol is a natural compound solated from Schisandra rubriflora; UGT2B7 UDP-glucuronosyltransferases inhibitor.