accessibility menu, dialog, popup

UserName

CAS RN 690632-73-6

HNNBr

CAS RN 690632-73-6

IUPAC Name: 1-[(4-bromophenyl)methyl]-1,4-diazepane
Synonyms: 1-[(4-bromophenyl)methyl]-1,4-diazepane
Molecular Weight (g/mol): 269.19
Molecular Formula: C12H17BrN2
InChi Key: PUTBARDRYLXING-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CN2CCCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results