CAS RN 685-88-1
CAS RN 685-88-1
IUPAC Name:
1,3-diethyl 2-fluoropropanedioate
Synonyms:
1,3-diethyl 2-fluoropropanedioate
Molecular Weight (g/mol):
178.16
Molecular Formula:
C7H11FO4
InChi Key:
GOWQBFVDZPZZFA-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(F)C(=O)OCC
1
–
2
of
2
results
1
Diethyl fluoromalonate, 97%
CAS: 685-88-1 Molecular Formula: C7H11FO4 Molecular Weight (g/mol): 178.159 MDL Number: MFCD00009139 InChI Key: GOWQBFVDZPZZFA-UHFFFAOYSA-N Synonym: diethyl fluoromalonate,diethyl 2-fluoromalonate,fluoromalonic acid diethyl ester,1,3-diethyl 2-fluoropropanedioate,diethylfluoromalonate,2-fluoro-malonic acid diethyl ester,diethyl2-fluoromalonate,diethyl fluoropropanedioate,propanedioic acid, fluoro-, diethyl ester,malonic acid, fluoro-, diethyl ester PubChem CID: 12702 IUPAC Name: diethyl 2-fluoropropanedioate SMILES: CCOC(=O)C(C(=O)OCC)F