CAS RN 6779-87-9
CAS RN 6779-87-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7,8-Dihydro-L-biopterin, MedChemExpress
MedChemExpress 7,8-Dihydro-L-biopterin is an oxidation product of tetrahydrobiopterin.