accessibility menu, dialog, popup

UserName

CAS RN 67698-61-7

CH3OO

CAS RN 67698-61-7

IUPAC Name: 3-propoxybenzaldehyde
Synonyms: 3-propoxybenzaldehyde
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: GFFJBSXKNZDYOJ-UHFFFAOYSA-N
SMILES: CCCOC1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results