CAS RN 67605-85-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-butyryl-L-Homoserine lactone, TRC
CAS: 67605-85-0 Molecular Formula: C8H13NO3 Molecular Weight (g/mol): 171.19 Synonym: N-[(3S)-Tetrahydro-2-oxo-3-furanyl]butanamide,N-Butanoyl-L-homoserine lactone,N-Butyryl-L-homoserine lactone,PAI,(S)-N-(Tetrahydro-2-oxo-3-furanyl)-butanamide IUPAC Name: N-[(3S)-2-oxooxolan-3-yl]butanamide SMILES: CCCC(=O)N[C@H]1CCOC1=O