accessibility menu, dialog, popup

UserName

CAS RN 675831-64-8

OHNN

CAS RN 675831-64-8

IUPAC Name: 3-(3-oxocyclopentyl)-1H-indole-5-carbonitrile
Synonyms: 3-(3-oxocyclopentyl)-1H-indole-5-carbonitrile
Molecular Weight (g/mol): 224.26
Molecular Formula: C14H12N2O
InChi Key: PCCIEBVCPXEIFS-UHFFFAOYNA-N
SMILES: O=C1CCC(C1)C1=CNC2=CC=C(C=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results