accessibility menu, dialog, popup

UserName

ONOOO

CAS RN 67565-48-4

IUPAC Name: 4-[(4-nitrophenyl)methoxy]benzaldehyde
Synonyms: 4-[(4-nitrophenyl)methoxy]benzaldehyde
Molecular Weight (g/mol): 257.25
Molecular Formula: C14H11NO4
InChi Key: RIYUFYPNHRLRON-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(COC2=CC=C(C=O)C=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results