CAS RN 67562-39-4
IUPAC Name:
3,4,5,6,10,11,12-heptachloro-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms:
3,4,5,6,10,11,12-heptachloro-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol):
409.29
Molecular Formula:
C12HCl7O
InChi Key:
WDMKCPIVJOGHBF-UHFFFAOYSA-N
SMILES:
ClC1=CC2=C(OC3=C2C(Cl)=C(Cl)C(Cl)=C3Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
1,2,3,4,6,7,8-Heptachlorodibenzofuran, TRC
CAS: 67562-39-4 Molecular Formula: C12 H Cl7 O Molecular Weight (g/mol): 409.31 Synonym: Dibenzofuran, 1,2,3,4,6,7,8-heptachloro-,1,2,3,4,6,7,8-Heptachlorodibenzofuran,1,2,3,4,6,7,8-HpCDF,1234678-HpCDF,F 131,HpCDF,PCDF 131,1,2,3,4,6,7,8-HeptaCDF,1234678-HeptaCDF,1234678-HpCDF,1234678-Heptachlorodibenzofuran IUPAC Name: 1,2,3,4,6,7,8-heptachlorodibenzofuran SMILES: Clc1cc2c(oc3c(Cl)c(Cl)c(Cl)c(Cl)c23)c(Cl)c1Cl