CAS RN 67400-25-3
CAS RN 67400-25-3
IUPAC Name:
3-bromo-5-nitro-2H-indazole
Synonyms:
3-bromo-5-nitro-2H-indazole
Molecular Weight (g/mol):
242.03
Molecular Formula:
C7H4BrN3O2
InChi Key:
SYTGRKBQTUKOAI-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC2=C(Br)NN=C2C=C1
1
–
1
of
1
results
1
3-Bromo-5-nitro-1H-indazole, 95%, Thermo Scientific™
CAS: 67400-25-3 Molecular Formula: C7H4BrN3O2 Molecular Weight (g/mol): 242.03 MDL Number: MFCD07367964 InChI Key: SYTGRKBQTUKOAI-UHFFFAOYSA-N Synonym: 3-bromo-5-nitro-1h-indazole,3-bromo-5-nitroindazole,5-nitro-3-bromoindazole,1h-indazole, 3-bromo-5-nitro,1h-indazole,3-bromo-5-nitro,5-nitro-3-bromo-indazole,pubchem20224,3-bromo-5-nitro1h-indazole,ksc497g0j,3-bromo-5-nitro 1h-indazole PubChem CID: 640614 IUPAC Name: 3-bromo-5-nitro-2H-indazole SMILES: [O-][N+](=O)C1=CC2=C(Br)NN=C2C=C1