CAS RN 67292-36-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]cobalt(II), 98%, Thermo Scientific™
CAS: 67292-36-8 Molecular Formula: C34H28Cl2CoFeP2 Molecular Weight (g/mol): 684.22 MDL Number: MFCD03427161 Synonym: [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride; CoCl2(dppf) IUPAC Name: [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride SMILES: [Fe].Cl[Co]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1