CAS RN 67292-26-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
[1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten, 98%, Thermo Scientific™
CAS: 67292-26-6 Molecular Formula: C38H28FeO4P2W Molecular Weight (g/mol): 850.27 MDL Number: MFCD03427160 IUPAC Name: [1,1'-Bis(diphenylphosphino)ferrocene]tetracarbonyltungsten SMILES: [Fe].[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1