CAS RN 67-07-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Creatine Phosphate (Phosphocreatine), >85%, TRC
CAS: 67-07-2 Molecular Formula: C4 H10 N3 O5 P Molecular Weight (g/mol): 211.11 Synonym: Glycine, N-[imino(phosphonoamino)methyl]-N-methyl-,Sarcosine, N-(phosphonoamidino)- (8CI),N-[Imino(phosphonoamino)methyl]-N-methylglycine,Creatine phosphate,Creatinephosphoric acid,N-(Phosphonoamidino)sarcosine,N-Phosphorocreatine,N-Phosphorylcreatine,Phosphocreatine,Phosphorylcreatine IUPAC Name: 2-[methyl-(N-phosphonocarbamimidoyl)amino]acetic acid SMILES: CN(CC(=O)O)C(=N)NP(=O)(O)O