accessibility menu, dialog, popup

UserName

CAS RN 6654-36-0

H3COOO

CAS RN 6654-36-0

IUPAC Name: methyl 6-oxohexanoate
Synonyms: methyl 6-oxohexanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: FDNFXHCDOASWAY-UHFFFAOYSA-N
SMILES: COC(=O)CCCCC=O
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results