CAS RN 6626-40-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7,10-Dichloro-2-methoxybenzo[b][1,5]naphthyridine, TRC
CAS: 6626-40-0 Molecular Formula: C13H8Cl2N2O Molecular Weight (g/mol): 279.12 Synonym: 7,10-Dichloro-2-methoxy-pyrido[3,2-b]quinoline,7,10-Dichloro-2-methoxypyrido[3,2-b]quinoline,NSC 59862 SMILES: COc1ccc2nc3cc(Cl)ccc3c(Cl)c2n1