accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 6609-57-0

IUPAC Name: 2-ethoxybenzonitrile
Synonyms: 2-ethoxybenzonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: DXTLCLWOCYLDHL-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)