accessibility menu, dialog, popup

UserName

ClClClClClKKRh

CAS RN 65980-75-8

IUPAC Name: rhodium(3+) dipotassium pentachloride
Synonyms: rhodium(3+) dipotassium pentachloride
Molecular Weight (g/mol): 358.35
Molecular Formula: Cl5K2Rh
InChi Key: YEPORAXCNSGGEV-UHFFFAOYSA-I
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Rh+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results