accessibility menu, dialog, popup

UserName

CH3SNH2NBr

CAS RN 65705-44-4

IUPAC Name: 4-(4-bromophenyl)-5-methyl-1,3-thiazol-2-amine
Synonyms: 4-(4-bromophenyl)-5-methyl-1,3-thiazol-2-amine
Molecular Weight (g/mol): 269.16
Molecular Formula: C10H9BrN2S
InChi Key: ULSZADQFOHESKR-UHFFFAOYSA-N
SMILES: CC1=C(N=C(N)S1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results