CAS RN 65567-96-6
IUPAC Name:
lithium(1+) potassium dichloride
Synonyms:
lithium(1+) potassium dichloride
Molecular Weight (g/mol):
116.94
Molecular Formula:
Cl2KLi
InChi Key:
HCQWRNRRURULEY-UHFFFAOYSA-L
SMILES:
[Li+].[Cl-].[Cl-].[K+]
1
–
1
of
1
results
1
Lithium chloride-potassium chloride
CAS: 65567-96-6 Molecular Formula: Cl2KLi Molecular Weight (g/mol): 116.938 MDL Number: MFCD00144623 InChI Key: HCQWRNRRURULEY-UHFFFAOYSA-L Synonym: lithium chloride-potassium chloride,acmc-20akmw,lithium potassium chloride,lithium potassium chloride 1/1/2,lithium chloride/potassium chloride eutectic,lithium 1+ ion potassium dichloride,lithium chloride/potassium chloride eutectic, anhydrous, beads trace metals basis PubChem CID: 16213218 IUPAC Name: lithium;potassium;dichloride SMILES: [Li+].[Cl-].[Cl-].[K+]