CAS RN 64790-15-4
CAS RN 64790-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress Glucagon (19-29), human
MedChemExpress Glucagon (19-29), human is a potent and efficient inhibitor of insulin secretion.
2
MedChemExpress Glucagon (19-29), human
MedChemExpress Glucagon (19-29), human is a potent and efficient inhibitor of insulin secretion.
3
MedChemExpress Glucagon (19-29), human
MedChemExpress Glucagon (19-29), human is a potent and efficient inhibitor of insulin secretion.